Order Entry
Slovenia
ContactUsLinkComponent
Biotinylated Mouse IL-15, Fc,Avitag (IL5-M82F3) is expressed from human 293 cells (HEK293). It contains AA Asn 49 - Ser 162 (Accessi on P48346-1). 1 * 200 µG
Catalog # ACRBIL5-M82F3200UG
Supplier:  ACRO Biosystems
Biotinylated Mouse IL-15, Fc,Avitag (IL5-M82F3) is expressed from human 293 cells (HEK293). It contains AA Asn 49 - Ser 162 (Accessi on P48346-1). 1 * 200 µG
Catalog # ACRBIL5-M82F3200UG
Supplier:  ACRO Biosystems
Frequently Bought Together