Order Entry
Slovenia
ContactUsLinkComponent
Human Complement C5 (w917s), His Tag (CO5-H52H7) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Cys 1676 (Acces sion P01031-1 (W917S)). 1 * 1 mg
Catalog # ACRBCO5-H52H7-1MG
Supplier:  ACRO Biosystems
Human Complement C5 (w917s), His Tag (CO5-H52H7) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Cys 1676 (Acces sion P01031-1 (W917S)). 1 * 1 mg
Catalog # ACRBCO5-H52H7-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together