Order Entry
Slovenia
Orders LinkContactUsLinkComponent
Human recombinant complement factor D (from HEK293 cells)

Human recombinant complement factor D (from HEK293 cells)

1 mg
Catalog # ACRBCFD-H52H8-1MG
Supplier:   ACRO Biosystems
Human recombinant complement factor D (from HEK293 cells)
Catalog # ACRBCFD-H52H8-1MG
Supplier:  ACRO Biosystems
CAS Number:  
Human recombinant complement factor D (from HEK293 cells)

1 mg

CAS Number:  
Price
2 650,00 €
Log in for availability
1 * 1 mg
Product Details & Documents

Human Complement Factor D / CFD Protein, His Tag (active enzyme), ACROBiosystems.

Specifications

Specifications

  • Pk:
    1 mg
  • Protein/peptide name:
    Complement Factor D
  • Protein synonyms:
    PFD|Complement factor D|CFD|DF|Adipsin
  • Purity:
    >90% as determined by SDS-PAGE.
  • Endotoxin level:
    Low
  • Source:
    HEK293 cells
  • Molecular weight:
    26,3 kDa
  • Shipping temperature:
    RT
  • Cat. No.:
    ACRBCFD-H52H8-1MG
  • Protein/peptide type:
    Recombinant
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Storage conditions:
    –20 °C
  • Gene ID:
    P00746-1
  • Formulation:
    Lyophilised from 0.22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH8.0. Normally glycerol is added as protectant before Lyophilisation.
  • Conjugation:
    Unconjugated
  • Tag sequence:
    His Tag (active enzyme)
  • Species:
    Human

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.