Order Entry
Slovenia
ContactUsLinkComponent
Biotinylated Human VEGF R1, His,Avitag (VE1-H82E3) is expressed from human 293 cells (HEK293). It contains AA Ser 27 - Asn 756 (Acce ssion P17948-1). 1 * 200 µG
Catalog # ACRBVE1-H82E3200UG
Supplier:  ACRO Biosystems
Biotinylated Human VEGF R1, His,Avitag (VE1-H82E3) is expressed from human 293 cells (HEK293). It contains AA Ser 27 - Asn 756 (Acce ssion P17948-1). 1 * 200 µG
Catalog # ACRBVE1-H82E3200UG
Supplier:  ACRO Biosystems
Frequently Bought Together