Order Entry
Slovenia
ContactUsLinkComponent
Human PD-L1, Strep Tag (PD1-H5282) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Arg 238 (Accession NP054862.1 ). 1 * 25 µG
Catalog # ACRBPD1-H5282-25UG
Supplier:  ACRO Biosystems
Human PD-L1, Strep Tag (PD1-H5282) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Arg 238 (Accession NP054862.1 ). 1 * 25 µG
Catalog # ACRBPD1-H5282-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together