Order Entry
Slovenia
ContactUsLinkComponent
Biotinylated Human B7-2, Fc,Avitag (CD6-H82F5) is expressed from human 293 cells (HEK293). It contains AA Leu 26 - Pro 247 (Accessio n AAH40261). 1 * 200 µG
Catalog # ACRBCD6-H82F5200UG
Supplier:  ACRO Biosystems
Biotinylated Human B7-2, Fc,Avitag (CD6-H82F5) is expressed from human 293 cells (HEK293). It contains AA Leu 26 - Pro 247 (Accessio n AAH40261). 1 * 200 µG
Catalog # ACRBCD6-H82F5200UG
Supplier:  ACRO Biosystems
Frequently Bought Together