Order Entry
Slovenia
ContactUsLinkComponent
Human BAFFR, Llama IgG2b Fc Tag, low endotoxin (BAR-H5258) is expressed from human 293 cells (HEK293). It contains AA Ser 7 - Ala 71 (Accession Q96RJ3-1). 1 * 100 µG
  ACRBBAR-H5258100UG
Human BAFFR, Llama IgG2b Fc Tag, low endotoxin (BAR-H5258) is expressed from human 293 cells (HEK293). It contains AA Ser 7 - Ala 71 (Accession Q96RJ3-1). 1 * 100 µG
  ACRBBAR-H5258100UG
Frequently Bought Together