Order Entry
Slovenia
ContactUsLinkComponent
Recombinat Protein,Rabbit Complement C5 Protein, His Tag,Rabbit Complement C5, His Tag (CO5-R52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Ala 1678 (Accession XP008271599.1).This protein carries a polyhistidine tag at the 1 * 100 µG
Catalog # ACRBCO5-R52H4100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Rabbit Complement C5 Protein, His Tag,Rabbit Complement C5, His Tag (CO5-R52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Ala 1678 (Accession XP008271599.1).This protein carries a polyhistidine tag at the 1 * 100 µG
Catalog # ACRBCO5-R52H4100UG
Supplier:  ACRO Biosystems
Frequently Bought Together