Recombinat Protein,Human CD209 DC-SIGN Protein, His Tag,Human CD209, His Tag (CD9-H5246) is expressed from human 293 cells (HEK293). It contains AA Gln 59 - Ala 404 (Accession Q9NNX6-1).This protein carries a polyhistidine tag at the N-terminus. The 1 * 1 mg
Catalog # ACRBCD9-H5246-1MG
Supplier: ACROBIOSYSTEMS
Frequently Bought Together