Human 4-1BB Ligand (71-254), His,Flag Tag (active trimer) (MALS verified) (41L-H52D4) is expressed from human 293 cells (HEK293). It contains AA Arg 71 - Glu 254 (Accession P41273-1) trimer Design. 1 * 100 µG
Catalog # ACRB41L-H52D4100UG
Supplier: ACRO Biosystems
Frequently Bought Together