Order Entry
Slovenia
ContactUsLinkComponent
Human 4-1BB Ligand (71-254), His,Flag Tag (active trimer) (MALS verified) (41L-H52D4) is expressed from human 293 cells (HEK293). It contains AA Arg 71 - Glu 254 (Accession P41273-1) trimer Design. 1 * 1 mg
Catalog # ACRB41L-H52D4-1MG
Supplier:  ACRO Biosystems
Human 4-1BB Ligand (71-254), His,Flag Tag (active trimer) (MALS verified) (41L-H52D4) is expressed from human 293 cells (HEK293). It contains AA Arg 71 - Glu 254 (Accession P41273-1) trimer Design. 1 * 1 mg
Catalog # ACRB41L-H52D4-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together