Order Entry
Slovenia
ContactUsLinkComponent
Cynomolgus CD3 epsilon, His Tag (CDE-C5226) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Asp 117 (Accession Q 95LI5-1). 1 * 100 µG
Catalog # ACRBCDE-C5226100UG
Supplier:  ACRO Biosystems
Cynomolgus CD3 epsilon, His Tag (CDE-C5226) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Asp 117 (Accession Q 95LI5-1). 1 * 100 µG
Catalog # ACRBCDE-C5226100UG
Supplier:  ACRO Biosystems
Frequently Bought Together