Order Entry
Slovenia
ContactUsLinkComponent
FITC-Labeled Human CD4, His Tag (CD4-HF2H7) is expressed from human 293 cells (HEK293). It contains AA Lys 26 - Trp 390 (Accession A AH25782). It is the FITC labeled form of Human CD4, His Tag (LE3-H5228). 1 * 200 µG
Catalog # ACRBCD4-HF2H7200UG
Supplier:  ACROBIOSYSTEMS
FITC-Labeled Human CD4, His Tag (CD4-HF2H7) is expressed from human 293 cells (HEK293). It contains AA Lys 26 - Trp 390 (Accession A AH25782). It is the FITC labeled form of Human CD4, His Tag (LE3-H5228). 1 * 200 µG
Catalog # ACRBCD4-HF2H7200UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together