Order Entry
Slovenia
ContactUsLinkComponent
Biotinylated Human IL-2, His,Avitag (IL2-H82E4) is expressed from human 293 cells (HEK293). It contains AA Ala 21 - Thr 153 (Accessi on P60568-1). 1 * 200 µG
Catalog # ACRBIL2-H82E4200UG
Supplier:  ACRO Biosystems
Biotinylated Human IL-2, His,Avitag (IL2-H82E4) is expressed from human 293 cells (HEK293). It contains AA Ala 21 - Thr 153 (Accessi on P60568-1). 1 * 200 µG
Catalog # ACRBIL2-H82E4200UG
Supplier:  ACRO Biosystems
Frequently Bought Together