Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Harnstoff FS, (4 x 20 ml R1, 1 x 20 ml R2) 1 * 1 KIT
Catalog # DIAS131019910021
Supplier:
DIAGNOSTIC SYSTEMS DIASYS
Expand image
Harnstoff FS, (4 x 20 ml R1, 1 x 20 ml R2) 1 * 1 KIT
Catalog # DIAS131019910021
Supplier:
DIAGNOSTIC SYSTEMS DIASYS
Frequently Bought Together
Hydrogen peroxide 6% (w/v) (20 volumes), GPR RECTAPUR®
Supplier: VWR Chemicals
Ammonia solution 25%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Fast Red RC Salt, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Test kits, photometric, COD, LCK cuvette tests
Supplier: Hach