Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
15ML MICROSPH.SIZE STDS 5.0µM 1 * 15 mL
Catalog # MSPP4205A
Supplier:
MARKET SOURCE PART PROCESS
Expand image
15ML MICROSPH.SIZE STDS 5.0µM 1 * 15 mL
Catalog # MSPP4205A
Supplier:
MARKET SOURCE PART PROCESS
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
15 ML MICROSPH. SIZE STDS. 25 µM 1 * 15 mL
Supplier: MARKET SOURCE PART PROCESS
COUNT-CAL PrecisionStandard 2µm, 6x25ml 1 * 150 mL
Supplier: MARKET SOURCE PART PROCESS