Order Entry
Slovenia
ContactUsLinkComponent
MULTI COMPONENT STANDARD 2 components 5000mg/l each of Chloromethane [CAS:74-87-3] Chloroethane [CAS:75-00-3] in Carbon disul fide . Duree de conservation : 12 mois. Store in a refrigerator at temperatures between 2C to 8C 1 * 1 mL
Catalog # CMPLRD0191121.6
MULTI COMPONENT STANDARD 2 components 5000mg/l each of Chloromethane [CAS:74-87-3] Chloroethane [CAS:75-00-3] in Carbon disul fide . Duree de conservation : 12 mois. Store in a refrigerator at temperatures between 2C to 8C 1 * 1 mL
Catalog # CMPLRD0191121.6
Frequently Bought Together