Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
KONTES FLEX-COLUMN Chromatography Columns, Kimble Chase, Preparative, 74 mL, 25 x 150 mm 1 * 5 items
Catalog # SCERSP420400-2515
Supplier:
DWK Life Sciences
Expand image
KONTES FLEX-COLUMN Chromatography Columns, Kimble Chase, Preparative, 74 mL, 25 x 150 mm 1 * 5 items
Catalog # SCERSP420400-2515
Supplier:
DWK Life Sciences
Frequently Bought Together
VWR®, Beakers, Low Form
Supplier: VWR Collection
VWR®, Pasteur Pipettes
Supplier: VWR Collection
VWR®, Laboratory Bottles, Narrow Neck, with Screw Cap
Supplier: VWR Collection
VWR®, Beakers, Low Form, Heavy-Duty
Supplier: VWR Collection
VWR® Flasks, Round Bottom, with Standard Ground joint
Supplier: VWR Collection
VWR® Erlenmeyer Flasks, with Standard Ground Joint
Supplier: VWR Collection
Laboratory bottles, narrow neck, with screw cap, DURAN®
Supplier: DWK Life Sciences
VWR®, Erlenmeyer Flasks
Supplier: VWR Collection
Screw caps for laboratory bottles, DURAN®
Supplier: DWK Life Sciences
Masterflex® Luer Stopcock Fittings, Polycarbonate, Avantor®
Supplier: Avantor Fluid Handling
Screw caps for laboratory bottles, DURAN®
Supplier: DWK Life Sciences