Order Entry
Slovenia
ContactUsLinkComponent
Mounting Kit with rubber element - Simple mounting solution for tanks and hoppers - Material: mild steel, chromated - Permissible horizontal displacement: 2.5 mm - for load cells up to 2.268kg 1 * 1 ST
Catalog # IBSAPR92/00N
Supplier:  IBS Stock Items
Mounting Kit with rubber element - Simple mounting solution for tanks and hoppers - Material: mild steel, chromated - Permissible horizontal displacement: 2.5 mm - for load cells up to 2.268kg 1 * 1 ST
Catalog # IBSAPR92/00N
Supplier:  IBS Stock Items
Frequently Bought Together