Ready-to-use cloths for disinfecting and cleaning surfaces of all kinds. 1 * 10 items
Catalog # SCUE70002034
Supplier: SCHUELKE MAYR
Ready-to-use cloths for disinfecting and cleaning surfaces of all kinds. 1 * 10 items
Catalog # SCUE70002034
Supplier: SCHUELKE MAYR
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
Autoclave tapes
Supplier: PRECISION DYNAMICS TIMEM
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Neutralising agent for the automated cleaning of glassware, neodisher® Z
Supplier: Dr. Weigert
Autoclavable bags, transparent, Clavies®, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Bottles, wide neck, with screw cap, Nalgene®
Supplier: Thermo Fisher Scientific
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
Microtubes, with thread
Supplier: SARSTEDT
Disinfectant solutions, Bacillol AF
Supplier: BODE CHEMIE