Order Entry
Slovenia
ContactUsLinkComponent
GreatSmart RP18 Column 100x4.6mm 3u 120A 1 * 1 items
Catalog # MAIS5141753
Supplier:  DR MAISCH
GreatSmart RP18 Column 100x4.6mm 3u 120A 1 * 1 items
Catalog # MAIS5141753
Supplier:  DR MAISCH
Frequently Bought Together