Order Entry
Slovenia
ContactUsLinkComponent
EGE IGFW 010 GSP/15 ******************** Induktiver Sensor M30x1,5x91, PTFE, Sn 10 b, 10...30 VDC, PNP-Schließer, -25...+120 °C, Kabel 15 m FEP 3x0,34 1 * 1 ST
Catalog # EGEE31440/15
EGE IGFW 010 GSP/15 ******************** Induktiver Sensor M30x1,5x91, PTFE, Sn 10 b, 10...30 VDC, PNP-Schließer, -25...+120 °C, Kabel 15 m FEP 3x0,34 1 * 1 ST
Catalog # EGEE31440/15
Frequently Bought Together