Order Entry
Slovenia
ContactUsLinkComponent
MSC2-C-53432220 PolyGard®2 Multi-Sensor-Controller MSC2 (MSC2-C-53432220) PolyGard®2 Multi-Sensor-Controller MSC2 Gehäuse Typ C 130 x 130 x 75 90-240 V AC / 24 V DC, 5 VA 3 x Alarmrelais Summer & Status-LED (rot, gelb, grün) Analog-Ausgang & RS 485 m 1 * 1 ST
Catalog # SNSTMSC2-C-5343222
MSC2-C-53432220 PolyGard®2 Multi-Sensor-Controller MSC2 (MSC2-C-53432220) PolyGard®2 Multi-Sensor-Controller MSC2 Gehäuse Typ C 130 x 130 x 75 90-240 V AC / 24 V DC, 5 VA 3 x Alarmrelais Summer & Status-LED (rot, gelb, grün) Analog-Ausgang & RS 485 m 1 * 1 ST
Catalog # SNSTMSC2-C-5343222
Frequently Bought Together