Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
UREA AGAR BASE (CHRISTENSEN) 1 * 500 g
Catalog # HIMEM112S-500G
Supplier:
HIMEDIA
Expand image
UREA AGAR BASE (CHRISTENSEN) 1 * 500 g
Catalog # HIMEM112S-500G
Supplier:
HIMEDIA
Frequently Bought Together
NBB®-B, Broth
Supplier: DOEHLER GROUP
SRK® Surfaces and Equipment Microbial Testing Devices
Supplier: Copan
VWR® Dehydrated Culture Media, Powdered
Supplier: VWR Chemicals
NBB®-A, Agar
Supplier: DOEHLER GROUP
VWR®, Contact slides
Supplier: VWR Chemicals
VWR® Ready to Use Media Plates (90 mm), Irradiated
Supplier: VWR Chemicals
VWR® Culture Media, Ready To Use, In Plates
Supplier: VWR Chemicals
Pepsin
Supplier: PanReac AppliChem
VWR® Contact Plates, Blister Packed, Irradiated
Supplier: VWR Chemicals