Order Entry
Slovenia
ContactUsLinkComponent
Stopper with platinum wire and platinum gauze, spare part for the determination of sulphur content in hydrocarbons, Schöninger 1 * 1 items
Catalog # WITG2.505.501
Supplier:  witeg Labortechnik
Stopper with platinum wire and platinum gauze, spare part for the determination of sulphur content in hydrocarbons, Schöninger 1 * 1 items
Catalog # WITG2.505.501
Supplier:  witeg Labortechnik
Frequently Bought Together