Biological indicators, strips
STRIP CONTROL GST E6: Paper strips inoculated with spores of Geobacillus stearothermophilus (ATCC® 7953) (1 - 5×106 CFU/strip), 100 strips
Biological indicators for validating the sterilisation using steam sterilisation (autoclaves).
- Description:STRIP CONTROL GST E6: Paper strips inoculated with spores of Geobacillus stearothermophilus (ATCC® 7953) (1 - 5×106 CFU/strip), 100 strips