Sartorius Cubis MCE20201S-2S00-0 Electronic High-Capacity Presicion Balance with user interface MCE with large, high-contrast liquid -crystal , weighing capacity 20200 g, readability 100 mg. 1 * 1 items
Catalog # 611-4760
Supplier: Sartorius Balances
Frequently Bought Together