Order Entry
Slovenia
ContactUsLinkComponent
TLC Glass Plates from Merck KGaA, Darmstadt, Germany, L x W 10 cm x 20 cm, silica gel 60 matrix, binder, Polymeric, fluorescent indi cator: no 1 * 1 Each
Catalog # SIALZ292966-1PAK
Supplier:  Merck
TLC Glass Plates from Merck KGaA, Darmstadt, Germany, L x W 10 cm x 20 cm, silica gel 60 matrix, binder, Polymeric, fluorescent indi cator: no 1 * 1 Each
Catalog # SIALZ292966-1PAK
Supplier:  Merck
Frequently Bought Together