Order Entry
Slovenia
ContactUsLinkComponent
Maxi 2-Desiccator Black, Aluminium frame, panels made of black acrylic glass prevent light incidence and protect against ultraviolet radiation, high-gloss surfaces, two compartments and two doors, including four shelves made of stainless steel, two y 1 * 1 items
Catalog # BOHLV1937-07
Supplier:  Bohlender
undefined
Maxi 2-Desiccator Black, Aluminium frame, panels made of black acrylic glass prevent light incidence and protect against ultraviolet radiation, high-gloss surfaces, two compartments and two doors, including four shelves made of stainless steel, two y 1 * 1 items
Catalog # BOHLV1937-07
Supplier:  Bohlender
Frequently Bought Together