Order Entry
Slovenia
ContactUsLinkComponent
An ELISA kit for the detection of TARC (Rat). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 18.75pg/ml. 1 * 1 KIT
  GENOIT7692
An ELISA kit for the detection of TARC (Rat). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 18.75pg/ml. 1 * 1 KIT
  GENOIT7692
Frequently Bought Together