Order Entry
Slovenia
ContactUsLinkComponent
ECH-Agarose contains carboxyl functional group for coupling of affinity ligands via their amine groups for affinity chromatography. ECH-Agarose is prepared by condensation of Carboxyl Coupling Resin (Cat. # 786-797) with succinic anhydride, resulting 1 * 50 mL
Catalog # GENO786-1224
Supplier:  G-Biosciences
ECH-Agarose contains carboxyl functional group for coupling of affinity ligands via their amine groups for affinity chromatography. ECH-Agarose is prepared by condensation of Carboxyl Coupling Resin (Cat. # 786-797) with succinic anhydride, resulting 1 * 50 mL
Catalog # GENO786-1224
Supplier:  G-Biosciences
Frequently Bought Together