0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Slovenia
ContactUsLinkComponent
[EN]ASTM D4815 QUANTITATIVE PEAK MIX 5X1
EHERGS09000186
:
EHRENSTORFER
[EN]ASTM D4815 QUANTITATIVE PEAK MIX 5X1
EHERGS09000186
:
EHRENSTORFER
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
: VWR Chemicals
pH buffers, ready to use
: WTW
pH buffer solutions
: Mettler - Toledo
VWR®, Conductivity standard solutions
: VWR Chemicals
Buffer solutions in sachets
: Mettler - Toledo