BsaI-HFv2 Menge/Units: 5.000 units - Lager/Store at: -20C 1 * 1 items
Catalog # NEWER3733L
Supplier: NEW ENGLAND BIOLABS
BsaI-HFv2 Menge/Units: 5.000 units - Lager/Store at: -20C 1 * 1 items
Catalog # NEWER3733L
Supplier: NEW ENGLAND BIOLABS
Frequently Bought Together
CryoBoxes with hinged lid, for cryovials, 1 to 5 ml, TruCool®
Supplier: BIOCISION
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF
Centrifuge tubes, 15 ml, Corning®
Supplier: Corning
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection
Multiwell cell culture plates, Falcon®
Supplier: Corning
Microtube and test tube racks, floating
Supplier: HEATHROW
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection
Cryoboxes, B69/B50/B80/B99-Series
Supplier: GLW
Serological pipettes, Stripette™
Supplier: Corning
Peqlab peqGOLD, Agarose, Universal
Supplier: VWR Chemicals
VWR® Blender Bags
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Spreaders, L-Shaped, Standard Line
Supplier: VWR Collection
VWR® Multiwell Cell Culture Plates
Supplier: VWR Collection
Vented QTray with cover, polystyrene, 240 x 240 x 20 mm 20 per case 1 * 20 items
Supplier: Molecular Devices
DNA ScreenTape Analysis (ScreenTape and Reagents)
Supplier: AGILENT
Genomic DNA ScreenTape Analysis (ScreenTape and Reagents)
Supplier: AGILENT
BL21(DE3) COMP. E.COLI 6 TUBES
Supplier: NEW ENGLAND BIOLABS
Q5 HS HF 2X MASTER MIX-100 RXNS
Supplier: NEW ENGLAND BIOLABS