Beckman Cell Holder for BioMate 160. Specially engineered microcell holder for Beckman style short microcells. Includes integrat ed mask to ensure that only the sample is illuminated by the beam. Also compatible with GENESYS 140, 150, 180. 1 * 1 items
Catalog # 634-1094
Supplier: Thermo Fisher Scientific
Frequently Bought Together