0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Slovenia
ContactUsLinkComponent
Conductivity Sensor: Single-use, nonsterile, PS, 1/2” HB 1 * 1 items
SPECACCS-12HB
:
Spectrum Laboratories
Conductivity Sensor: Single-use, nonsterile, PS, 1/2” HB 1 * 1 items
SPECACCS-12HB
:
Spectrum Laboratories
Frequently Bought Together
Filtration capsules, Sartopore® 2 XLG
: Sartorius
STORAGE SET FOR ELECTRODES SET 2 1 * 1 items
: Roth Carl
Dissolved oxygen sensors for industrial applications, OxyFerm FDA
: HAMILTON BONADUZ
Flathead mop covers and hardware, Slimline™
: MICRONOVA