Order Entry
Slovenia
ContactUsLinkComponent
Volume Range: 100 ìL – 1 mL Increment (electric): 1 ìL Volume: 1 mL Accuracy ±: 1 % / 1 mL 1 % / 0.5 mL 2.5 % / 0.1 mL Precision ±: 0.4 % / 1 mL 0.4 % / 0.5 mL 1.25 % / 0.1 mL : LiteTouch System LTS High-throughput Pipette Semi-automated 96-w 1 * 1 items
Catalog # RAIN30296707
Supplier:  RAININ
Volume Range: 100 ìL – 1 mL Increment (electric): 1 ìL Volume: 1 mL Accuracy ±: 1 % / 1 mL 1 % / 0.5 mL 2.5 % / 0.1 mL Precision ±: 0.4 % / 1 mL 0.4 % / 0.5 mL 1.25 % / 0.1 mL : LiteTouch System LTS High-throughput Pipette Semi-automated 96-w 1 * 1 items
Catalog # RAIN30296707
Supplier:  RAININ
Frequently Bought Together