Order Entry
Slovenia
ContactUsLinkComponent
Agilent J+W DB-5 Intuvo GC column module, 30 m, 0.32 mm, 0.25 µm 1 * 1 items
Catalog # HEWL123-5032-INT
Supplier:  Agilent
undefined
Agilent J+W DB-5 Intuvo GC column module, 30 m, 0.32 mm, 0.25 µm 1 * 1 items
Catalog # HEWL123-5032-INT
Supplier:  Agilent
Frequently Bought Together