PROTEINASE K SOLUTION 5ML (20MG/ML) GENOMIC PCR GRADE 1 * 5 mL
Catalog # VIAG501-PK
Supplier: VIAGEN BIOTECH
PROTEINASE K SOLUTION 5ML (20MG/ML) GENOMIC PCR GRADE 1 * 5 mL
Catalog # VIAG501-PK
Supplier: VIAGEN BIOTECH
Frequently Bought Together
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
VWR® Water Sampling Bottles, with Tamper Evident Cap
Supplier: VWR Collection
VWR®, Conical Sample Containers, with Snap-On Lids
Supplier: VWR Collection
VWR® Blender Bags
Supplier: VWR Collection
Centrifuge tubes
Supplier: Brand
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Sampling Spoons
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection