2-Fluorocyclohexanone 96%
Bottle, 250 mg
Catalog # APOSPC201281-250MG
Supplier: Apollo Scientific
2-Fluorocyclohexanone 96%
Catalog # APOSPC201281-250MG
Supplier: Apollo Scientific
CAS Number: 694-82-6
Jump to:
Product Details & Documents
- CAS Number:694-82-6
- Formula:C₆H₉FO
- MDL Number:MFCD01076476
- Molecular Weight:116.14 g/mol
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
- Pk:250 mg



