Order Entry
Slovenia
ContactUsLinkComponent
Ciliary neurotrophic factor (CNTF) is a neurotrophic factor that promotes the survival of neuronal cell populations, neurite outgrow th, and neurotransmitter synthesis. CNTF also plays an important protective role during nervous system injury. 1 * 500 µG
Catalog # SHBT200-90AF-500UG
undefined
Ciliary neurotrophic factor (CNTF) is a neurotrophic factor that promotes the survival of neuronal cell populations, neurite outgrow th, and neurotransmitter synthesis. CNTF also plays an important protective role during nervous system injury. 1 * 500 µG
Catalog # SHBT200-90AF-500UG
Frequently Bought Together