Order Entry
Slovenia
ContactUsLinkComponent
Pipette calibration kit (hardware) for balances with a readability of 0.01 mg. The kit consists of a moisture trap and all required adapters 1 * 1 items
Catalog # SARBYCP06SQ
Supplier:  Sartorius Balances
Pipette calibration kit (hardware) for balances with a readability of 0.01 mg. The kit consists of a moisture trap and all required adapters 1 * 1 items
Catalog # SARBYCP06SQ
Supplier:  Sartorius Balances
Frequently Bought Together