Order Entry
Slovenia
ContactUsLinkComponent
marking the side of 0.5 to 50 ml tubes Includes: 4 x tube adaptors (4ti-0681, 4ti-0682, 4ti-0683 and 4ti- 0684), AC adaptor 18W 18VDC (input 100-240VAC), USB A-B cable, operation manual, TubeMarker™ software (on USB- Stick), 12 months warranty 1 * 1 items
Catalog # FTIT4TI-0680-1
Supplier:  4titude Ltd.
undefined
marking the side of 0.5 to 50 ml tubes Includes: 4 x tube adaptors (4ti-0681, 4ti-0682, 4ti-0683 and 4ti- 0684), AC adaptor 18W 18VDC (input 100-240VAC), USB A-B cable, operation manual, TubeMarker™ software (on USB- Stick), 12 months warranty 1 * 1 items
Catalog # FTIT4TI-0680-1
Supplier:  4titude Ltd.
Frequently Bought Together