Order Entry
Slovenia
ContactUsLinkComponent
Photometer PF-3 Drinking Water for evaluation of VISOCOLOR ECO tests and NANOCOLOR tube tests (Cl2, pH, F-, Fe, ClO2) incl. manual, 3 batteries and certificate in a box 1 * 1 SET
Catalog # MANA919343
Supplier:  MACHEREY-NAGEL
undefined
Photometer PF-3 Drinking Water for evaluation of VISOCOLOR ECO tests and NANOCOLOR tube tests (Cl2, pH, F-, Fe, ClO2) incl. manual, 3 batteries and certificate in a box 1 * 1 SET
Catalog # MANA919343
Supplier:  MACHEREY-NAGEL