Order Entry
Slovenia
ContactUsLinkComponent
NucleoSpin 8 PCR Clean-up (12 x 8) 12 x 8 preps for the purification of PCR products - NucleoSpin PCR Clean-up Binding Strips, MN Wa sh Plate, Rack of Tube Strips, Elution Plate U-bottom, buffers no dangerous goods according transport regulations 1 * 12 items
Catalog # MANA740668
Supplier:  MACHEREY-NAGEL
undefined
NucleoSpin 8 PCR Clean-up (12 x 8) 12 x 8 preps for the purification of PCR products - NucleoSpin PCR Clean-up Binding Strips, MN Wa sh Plate, Rack of Tube Strips, Elution Plate U-bottom, buffers no dangerous goods according transport regulations 1 * 12 items
Catalog # MANA740668
Supplier:  MACHEREY-NAGEL
Frequently Bought Together