Order Entry
Slovenia
ContactUsLinkComponent
NucleoSpin 8 Blood QuickPure (12x8) 12x 8 preps for the isolation of genomic DNA from blood - NucleoSpin Blood QuickPure Binding Str ips, Racks of Tube Strips, Self-adhering PE Foils, buffers, Proteinase K no dangerous goods according transport regu 1 * 12 items
Catalog # MANA740666
Supplier:  MACHEREY-NAGEL
undefined
NucleoSpin 8 Blood QuickPure (12x8) 12x 8 preps for the isolation of genomic DNA from blood - NucleoSpin Blood QuickPure Binding Str ips, Racks of Tube Strips, Self-adhering PE Foils, buffers, Proteinase K no dangerous goods according transport regu 1 * 12 items
Catalog # MANA740666
Supplier:  MACHEREY-NAGEL
Frequently Bought Together