NucleoSnap Plasmid Midi (50) 50 preps for the isolation of transfection-grade plasmid DNA - NucleoSnap Plasmid Columns, NucleoSpin P lasmid Filter Columns, buffers, RNase A __UN 1993 Flammable liquid, n.o.s. (2-propanol mixture) 3 1 * 50 items
Catalog # MANA740494.50
Supplier: MACHEREY-NAGEL
Frequently Bought Together