Order Entry
Slovenia
ContactUsLinkComponent
Therm. straight stem, similar DIN 16174, 0+300:2°C, cap. prismatic, yellow backed, milk glass scale, eco-friendly, green liquid, bio degradable, upper part 235x20,5mm, lower part 165x8mm, immersion 138mm, cork-glass-top finish 1 * 1 items
Catalog # AMART91296-EF
Supplier:  Amarell
Therm. straight stem, similar DIN 16174, 0+300:2°C, cap. prismatic, yellow backed, milk glass scale, eco-friendly, green liquid, bio degradable, upper part 235x20,5mm, lower part 165x8mm, immersion 138mm, cork-glass-top finish 1 * 1 items
Catalog # AMART91296-EF
Supplier:  Amarell
Frequently Bought Together