Order Entry
Slovenia
ContactUsLinkComponent
Flash point-Therm., similar Abel-Pensky, similar DIN, -30+30:0,5C, 57mm immersion, max. 280x9-10mm, milk glass scale, cap. prismati c, colourless, red special liquid, Richter-top finish, with swelling, spherical bulb, suitable for government verific 1 * 1 items
Catalog # AMARM230006-FL
Supplier:  Amarell
Flash point-Therm., similar Abel-Pensky, similar DIN, -30+30:0,5C, 57mm immersion, max. 280x9-10mm, milk glass scale, cap. prismati c, colourless, red special liquid, Richter-top finish, with swelling, spherical bulb, suitable for government verific 1 * 1 items
Catalog # AMARM230006-FL
Supplier:  Amarell
Frequently Bought Together