Order Entry
Slovenia
ContactUsLinkComponent
Flash point therm., similar Abel-Pensky, similar DIN 12785, 0+70:0,5C, 57mm immersion, max. 280x9-10mm, milk glass scale, cap. pri smatic, colourless, red special liquid, Richter-top finish, with swelling, spherical bulb, null
Catalog # AMARM230000-FL/VC
Supplier:  Amarell
Flash point therm., similar Abel-Pensky, similar DIN 12785, 0+70:0,5C, 57mm immersion, max. 280x9-10mm, milk glass scale, cap. pri smatic, colourless, red special liquid, Richter-top finish, with swelling, spherical bulb, null
Catalog # AMARM230000-FL/VC
Supplier:  Amarell
Frequently Bought Together