Order Entry
Slovenia
ContactUsLinkComponent
Cardanic suspension devicese for work with hydrometers, consist of: special holding device made of stainless steel with tripod and m easuring cylinder made of glass, height 450mm, diameter 55mm, capacity 1000ml, total height 600mm 1 * 1 items
Catalog # AMARH860910
Supplier:  Amarell
Cardanic suspension devicese for work with hydrometers, consist of: special holding device made of stainless steel with tripod and m easuring cylinder made of glass, height 450mm, diameter 55mm, capacity 1000ml, total height 600mm 1 * 1 items
Catalog # AMARH860910
Supplier:  Amarell
Frequently Bought Together